Learn More
Labconco™ WaterPro™ PS Water Polishing Station
Product Code. 15492212 Shop All Labconco Corporation Products
Change view
Click to view available options
Type:
PS and PS/UF Startup Kit
PS/HPLC and PS HPLC/UF Startup Kit
Model:
WaterPro PS HPLC/UF Hybrid
WaterPro PS and PS/UF
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Model Unit Size
15492212 PS and PS/UF Startup Kit WaterPro PS and PS/UF Each
15302104 PS/HPLC and PS HPLC/UF Startup Kit WaterPro PS HPLC/UF Hybrid Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15492212 Supplier Labconco™ Supplier No. 9047101
  • ONLINE EXCLUSIVE PRICE
    722.00 EUR
    691.46 EUR / Pack of 1
    Save 30.54 EUR
    4% Off
 Restricted Item
Estimated Shipment: 12-01-2027
WaterPro™ PS Water Polishing Station with filters, prefilters and cartridges for Labconco WaterPro PS and RO Water Purification Systems.
  • General Chemistry/UF Polishing Kit includes Activated Carbon Cartridge (1), polishing grade mixed bed Deionization Cartridges (3).
  • Kit is required for models 9000520, 9000521, 9000530, 9000531, 9000540, 9000541, 9000560, 9000561, 9000570, 9000571, 9000580, 9000581, 9000720, 9000721, 9000730, 9000731, 9000740, 9000741, 9000760, 9000761, 9000770, 9000771, 9000780 and 9000781.
Order Info
All models require Start-Up Kits, sold separately. Unit dimensions (including bowls) (L x W x H): 7 3/4 x 31 1/8 x 28 7/8 in. (20 x 79 x 73 cm). For 115 V 60 Hz, 230 V 50 Hz models also available through your Fisher Scientific™ Representative.
†Comes with valve to dispense Type I water and gun to dispense Type I, pyrogen-free water.
Specifications
Description WaterPro PS and PS/UF Startup Kit
Bacterial Content 0.001 CFU/mL
Daily Output 1 to 250 L
Endotoxin Concentration <0.001 EU/mL
Feedwater Source Reverse Osmosis Tap
Flow Rate 1.1 L/min.
Product Water Type Type 1
Resistivity 16 to 18.2 MΩ·cm
Total Organic Carbon <30 ppb
Type PS and PS/UF Startup Kit
For Use With (Equipment) WaterPro™ PS polishing station
Includes Activated Carbon cartridge (1) and polishing grade mixed bed deionization cartridges (3)
Model WaterPro PS and PS/UF
Weight (English) 9.5 lb.
Weight (Metric) 4.3 kg