Learn More
Miele Professional™ Water purification
Product Code. 10428922 Shop All Miele Products
Change view
Click to view available options
For Use With (Equipment):
Washer-disinfectors
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
10428922 Washer-disinfectors Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10428922 Supplier Miele Professional™ Supplier No. 3830350
  • ONLINE EXCLUSIVE PRICE
    475.00 EUR
    315.00 EUR / Pack of 1
    Save 160.00 EUR
    33% Off
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
Full demineralisation single use resin E 315 for the regeneration of full demineralisation cartridges
  • 2 bags each containing 10 litres
  • For empty cartridge LP 2800 and full demin. cartridge VE 2000/2800
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Washer-disinfectors
Type Water Pre-treatment