Learn More
Miele Professional™ Water purification
Product Code. 15420535 Shop All Miele Products
Change view
Click to view available options
For Use With (Equipment):
Washer-disinfectors
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15420535 Washer-disinfectors Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15420535 Supplier Miele Professional™ Supplier No. 10020100
  • ONLINE EXCLUSIVE PRICE
    122.00 EUR
    101.00 EUR / Pack of 1
    Save 21.00 EUR
    17% Off
 Restricted Item
Estimated Shipment: 19-10-2026
Conversion kit for two cartridges (UfZ) for the quick change to a full/partial demineralisation cartridge
2 x VA plug connection 3/4'' for installation of second cartridge
TRUSTED_SUSTAINABILITY
Specifications
Color (Overall) Stainless Steel
For Use With (Equipment) Washer-disinfectors
Type Water Pre-treatment
Material Stainless Steel
Net Weight (Metric) 0.15 kg