Learn More
Bryson Packaging™ Refuse Sacks
Product Code. 12463070
Change view
Click to view available options
Dimensions: 450L x 800W x 975mmH
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
12463070 Dimensions: 450L x 800W x 975mmH
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
You may only order up to the available stock. For more details call customer services on +358 9 8027 6280.
Product Code. 12463070 Supplier Bryson Packaging™ Supplier No. WDS650525X
  • ONLINE EXCLUSIVE PRICE
    1075.90 EUR
    52.77 EUR / Pack of 100
    Save 1023.13 EUR
    95% Off
Estimated Shipment: 22-12-2026
You may only order up to the available stock. For more details call customer services on +358 9 8027 6280.
An excellent option for refuse and trash removal from the laboratory. Bryson Packaging™ Refuse Sacks are manufactured from polyethylene.
  • Durable to prevent leaks
  • Provided in a packs of 100
Specifications
Product Type Waste Sack