Learn More
Cytiva Wash Valve
Product Code. 15209774 Shop All Cytiva Products
Change view
Click to view available options
For Use With (Equipment):
AKTA start Automated Protein Purification System
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15209774 AKTA start Automated Protein Purification System Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15209774 Supplier Cytiva Supplier No. 29023897
Estimated Shipment: 29-10-2026
For use with the AKTA start automated protein purification system. The wash valve is used to divert the flow path to the waste when needed.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Divert the flow path to the waste when needed
For Use With (Equipment) AKTA start Automated Protein Purification System