Learn More
Hughes Wall Mounted Indoor Unheated Safety Shower
Product Code. 17742348
Change view
Click to view available options
Mounting:
Wall
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mounting Unit Size
17742348 Wall Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17742348 Supplier Hughes Supplier No. HSHH0023GS
  • ONLINE EXCLUSIVE PRICE
    711.21 EUR
    493.97 EUR / Pack of 1
    Save 217.24 EUR
    30% Off
Estimated Shipment: 14-12-2026
Wall mounted unheated emergency safety shower, intended for use in environments where space is at a premium. This drench shower is suitable for use in all applications that don’t require an eyewash and are not subject to the possibility of freezing or overheating.
  • Wall mounted unheated emergency safety shower, intended for use in environments where space is at a premium
  • This drench shower is suitable for use in all applications that don't require an eyewash and are not subject to the possibility of freezing or overheating
Specifications
For Use With (Application) Indoor Applications
Material Stainless Steel
Mounting Wall
Product Type Safety Shower