Learn More
Hughes Wall Mounted Closed Bowl Emergency Eye/Face Wash
Product Code. 17762338
Change view
Click to view available options
Mounting:
Wall
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mounting Unit Size
17762338 Wall Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17762338 Supplier Hughes Supplier No. HEWS0045GV
  • ONLINE EXCLUSIVE PRICE
    558.01 EUR
    357.20 EUR / Pack of 1
    Save 200.81 EUR
    35% Off
Estimated Shipment: 17-12-2026
Wall mounted, plumbed-in emergency eye/face wash with closed ABS bowl.
  • Wall-mounted emergency eye/face wash with integral lid to protect the bowl and diffusers against dust and debris
  • This ANSI compliant, plumbed in eye/face wash is designed to be quick and simple to operate with high flow rate aerated diffusers for improved washing
Specifications
Color Green, White
Height (Metric) 36 cm
Mounting Wall
Width (Metric) 32 cm