Learn More
Emma Safety Footwear Vulcanus XD Safety Shoes
Product Code. 15998711
Change view
Click to view available options
Size:
39
40
41
42
43
44
45
46
47
48
49
Unit Size:
1 pair
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15998711 44 1 pair
15948711 39 1 pair
15958711 40 1 pair
15968711 41 1 pair
15978711 42 1 pair
15988711 43 1 pair
15908721 45 1 pair
15918721 46 1 pair
15928721 47 1 pair
15938721 48 1 pair
15948721 49 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Product Code. 15998711 Supplier Emma Safety Footwear Supplier No. MM54086844
  • ONLINE EXCLUSIVE PRICE
    159.91 EUR
    93.71 EUR / 1 pairs
    Save 66.20 EUR
    41% Off
Estimated Shipment: 24-03-2027
Welder's boot designed specifically for working with metal in extreme temperatures. Over-foot flap provides no entry for sparks or shards.
  • S3-Safety shoe with sole plate for wet work
  • Full grain leather construction
  • GripForce™ Duo Heat Resistant Outsole
  • Velcro over-foot flap
  • Sanitized
Specifications
Design Vulcanus XD
Material Leather
Size 44