Learn More
Vortioxetine Hydrobromide, TRC
Product Code. 30009227
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
50 mg
Unit Size:
10mg
2.5mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009227 50 mg 50mg
17961565 10 mg 10mg
17982955 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30009227 Supplier Toronto Research Chemicals Supplier No. TRCV76600050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 960203-27-4
Molecular Formula C18 H22 N2 S . Br H
Molecular Weight (g/mol) 379.358
Synonym Piperazine, 1-[2-[(2,4-dimethylphenyl)thio]phenyl]-, hydrobromide (1:1), 1-[2-[(2,4-Dimethylphenyl)thio]phenyl]piperazine hydrobromide, 1-[2-(2,4-Dimethylphenylsulfanyl)phenyl]piperazine hydrobromide, Lu AA 21004 hydrobromide, Vortioxetine hydrobromide
IUPAC Name 1-[2-(2,4-dimethylphenyl)sulfanylphenyl]piperazine;hydrobromide
SMILES Br.Cc1ccc(Sc2ccccc2N3CCNCC3)c(C)c1
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C18H22N2S.BrH/c1-14-7-8-17(15(2)13-14)21-18-6-4-3-5-16(18)20-11-9-19-10-12-20;/h3-8,13,19H,9-12H2,1-2H3;1H
Formula Weight 378.077
Percent Purity >95%
Chemical Name or Material Vortioxetine Hydrobromide
Grade HPLC