Learn More
Vortioxetine Hydrobromide (1.0 mg/mL in Methanol), TRC
Specifications
Specifications
| CAS | 960203-27-4 |
| Molecular Formula | C18 H22 N2 S . Br H |
| Quantity | 5 x 1 mL |
| Synonym | Piperazine, 1-[2-[(2,4-dimethylphenyl)thio]phenyl]-, hydrobromide (1:1), 1-[2-[(2,4-Dimethylphenyl)thio]phenyl]piperazine hydrobromide, 1-[2-(2,4-Dimethylphenylsulfanyl)phenyl]piperazine hydrobromide, Lu AA 21004 hydrobromide, Vortioxetine hydrobromide |
| InChI Formula | InChI=1S/C18H22N2S.BrH/c1-14-7-8-17(15(2)13-14)21-18-6-4-3-5-16(18)20-11-9-19-10-12-20;/h3-8,13,19H,9-12H2,1-2H3;1H |
| SMILES | Br.Cc1ccc(Sc2ccccc2N3CCNCC3)c(C)c1 |
| IUPAC Name | 1-[2-(2,4-dimethylphenyl)sulfanylphenyl]piperazine;hydrobromide |
| Molecular Weight (g/mol) | 379.358 |
| Formula Weight | 378.077 |
| Chemical Name or Material | Vortioxetine Hydrobromide |