Learn More
Scientific Industries SI™ Vortex-Genie™ 2
Product Code. 15547335 Shop All Ohaus Products
Change view
Click to view available options
Plug Type:
EU Plug
Model:
SI-0256
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Plug Type Model Unit Size
15547335 EU Plug SI-0256 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15547335 Supplier Scientific Industries SI™ Supplier No. 83041404
  • ONLINE EXCLUSIVE PRICE
    777.67 EUR
    386.34 EUR / Pack of 1
    Save 391.33 EUR
    50% Off
View all promotions
Estimated Shipment: 19-10-2026
Add to basket
Add to basket
Mixes gently, vigorously, and anywhere in between. Scientific Industries SI™ Vortex-Genie™ 2 speeds range from 600 to 3200rpm.
  • Variable speed for precise mixing
  • Broad range of attachments available for most tubes, plates and other containers
  • Hands-Free or Touch On control
  • May be used in cold rooms and incubators
TRUSTED_SUSTAINABILITY
Specifications
Speed Range 600 to 3200 rpm
Electrical Requirements 230 V, 60 Hz
Voltage 230 V
Product Type Vortex Mixer
Certifications/Compliance CE, ETL
Plug Type EU Plug
Frequency 60 Hz
Depth (Metric) 165 mm
Height (Metric) 165 mm
Width (Metric) 122 mm
Weight (Metric) 4 kg
Model SI-0256
Includes Pop-off cup, 76mm platform
Operating Modes Hands-free, Touch on
Warranty and Service Offering 2 Years