missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Von Willebrand Factor Antibody (VWF/1465), Novus Biologicals™
Mouse Monoclonal Antibody
290.00 EUR - 542.00 EUR
Specifications
| Antigen | vWF-A2 |
|---|---|
| Clone | VWF/1465 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 0.5 - 1.0 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunoprecipitation 0.5 - 1 ug/500ug, Immunohistochemistry-Paraffin 1:10-1:500, SDS-Page |
| Applications | Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18240030
|
Novus Biologicals
NBP2-53314-20UG |
20 μg |
290.00 EUR
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18233630
|
Novus Biologicals
NBP2-53314-100UG |
100 μg |
542.00 EUR
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Von Willebrand Factor Monoclonal specifically detects Von Willebrand Factor in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.Specifications
| vWF-A2 | |
| 0.2 mg/mL | |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human | |
| F8VWF, VWD, VWF von Willebrand factor | |
| VWF | |
| IgG1 | |
| Protein A or G purified | |
| 250 kDa |
| VWF/1465 | |
| Western Blot 0.5 - 1.0 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunoprecipitation 0.5 - 1 ug/500ug, Immunohistochemistry-Paraffin 1:10-1:500, SDS-Page | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
| 7450 | |
| Recombinant fragment of human vWF protein (aa1815-1939) (exact sequence is proprietary) | |
| Primary | |
| Store at 4C. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title