missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Von Willebrand Factor Antibody (VWF/1465), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-53314-20UG
This item is not returnable.
View return policy
Description
Von Willebrand Factor Monoclonal specifically detects Von Willebrand Factor in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.
Specifications
| vWF-A2 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Western Blot 0.5 - 1.0 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunoprecipitation 0.5 - 1 ug/500ug, Immunohistochemistry-Paraffin 1:10-1:500, SDS-Page | |
| F8VWF, VWD, VWF von Willebrand factor | |
| Mouse | |
| 250 kDa | |
| 20 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| VWF/1465 | |
| Unconjugated | |
| 10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
| VWF | |
| Recombinant fragment of human vWF protein (aa1815-1939) (exact sequence is proprietary) | |
| Protein A or G purified | |
| RUO | |
| 7450 | |
| Store at 4C. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction