Learn More
Hirschmann™ Volumetric Measuring Device Fitting
Product Code. 10293861
Change view
Click to view available options
For Use With (Equipment):
Schilling Fast-Flow Burettes, 15 - 50 mL
Schilling Fast-Flow Burettes, 5 - 10 mL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
10293861 Schilling Fast-Flow Burettes, 15 - 50 mL Each
10069770 Schilling Fast-Flow Burettes, 5 - 10 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10293861 Supplier Hirschmann™ Supplier No. 8900750
  • ONLINE EXCLUSIVE PRICE
    25.00 EUR
    22.30 EUR / Pack of 1
    Save 2.70 EUR
    10% Off
Estimated Shipment: 16-12-2026
Fitting for Dr. Schilling fast-flow burettes
  • For Schilling burettes
Specifications
For Use With (Equipment) Schilling Fast-Flow Burettes, 15 - 50 mL
No. per Pack 1 Each
Type Buret Fitting
Product Title
Select an issue
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privatlivspolitik.