Learn More
VOC Mixture 928 2000 μg/mL in Water, Dr. Ehrenstorfer
Specifications
Specifications
| CAS | 123-91-1, 71-23-8, 78-93-3, 591-78-6, 108-10-1, 67-64-1 |
| Quantity | 1 mL |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Water |