Learn More
VOC Mixture 561 1000 μg/mL in Methanol, Dr. Ehrenstorfer
Product Code. 30328008
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30328008 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30328008 Supplier Dr. Ehrenstorfer Supplier No. DREA50000561ME
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
CAS 71-43-2, 123-86-4, 100-41-4, 108-38-3, 544-76-3, 110-54-3
Quantity 1 mL
Certifications/Compliance ISO 17034
Solvent or Matrix Methanol