missing translation for 'onlineSavingsMsg'
Learn More

VOC-Mix 15 200 μg/mL in Methanol, Dr. Ehrenstorfer

Product Code. 30321396
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30321396 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30321396 Supplier Dr. Ehrenstorfer Supplier No. DREXA05000015ME
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

CAS 630-20-6, 71-55-6, 79-34-5, 79-00-5, 75-34-3, 75-35-4
Quantity 1 mL
Concentration 1,1,1,2-Tetrachloroethane 200 μg/mL∣1,1,1-Trichloroethane 200 μg/mL∣1,1,2,2-Tetrachloroethane 200 μg/mL∣1,1,2-Trichloroethane 200 μg/mL∣1,1-Dichloroethane 200 μg/mL∣1,1-Dichloroethene 200 μg/mL∣1,1-Dichloropropene 200 μg/mL∣1,2,3-Trichlorobenzene 200 μg/mL∣1,2,3-Trichloropropane 200 μg/mL∣1,2,4-Trichlorobenzene 200 μg/mL∣1,2,4-Trimethylbenzene 200 μg/mL∣1,2-Dibromo-3-chloropropane 200 μg/mL∣1,2-Dibromoethane 200 μg/mL∣1,2-Dichlorobenzene 200 μg/mL∣1,2-Dichloroethane 200 μg/mL∣1,2-Dichloropropane 200 μg/mL∣1,3,5-Trimethylbenzene 200 μg/mL∣1,3-Dichlorobenzene 200 μg/mL∣1,3-Dichloropropane 200 μg/mL∣1,4-Dichlorobenzene 200 μg/mL∣2,2-Dichloropropane 200 μg/mL∣2-Chlorotoluene 200 μg/mL∣4-Chlorotoluene 200 μg/mL∣4-Cymene 200 μg/mL∣Benzene 200 μg/mL∣Bromobenzene 200 μg/mL∣Bromochloromethane 200 μg/mL∣Bromodichloromethane 200 μg/mL∣Chlorobenzene 200 μg/mL∣Chloroform 200 μg/mL∣cis-1,2-Dichloroethene 200 μg/mL∣cis-1,3-Dichloropropene 200 μg/mL∣Dibromochloromethane 200 μg/mL∣Dibromomethane 200 μg/mL∣Dichloromethane (Methylenechloride) 200 μg/mL∣Ethylbenzene 200 μg/mL∣Hexachlorobutadiene 200 μg/mL∣Isopropylbenzene 200 μg/mL∣m-Xylene (1,3-Dimethylbenzene) 200 μg/mL∣Naphthalene 200 μg/mL∣n-Butylbenzene 200 μg/mL∣o-Xylene (1,2-Dimethylbenzene) 200 μg/mL∣Propylbenzene 200 μg/mL∣p-Xylene (1,4-Dimethylbenzene) 200 μg/mL∣sec-Butylbenzene 200 μg/mL∣Styrene 200 μg/mL∣tert-Butylbenzene 200 μg/mL∣Tetrachloroethene 200 μg/mL∣Tetrachloromethane 200 μg/mL∣Toluene 200 μg/mL∣trans-1,2-Dichloroethene 200 μg/mL∣trans-1,3-Dichloropropene 200 μg/mL∣Tribromomethane 200 μg/mL∣Trichloroethene 200 μg/mL
Certifications/Compliance ISO/IEC 17025
Solvent or Matrix Methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.