missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vitronectin Antibody (A18) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
199.00 EUR - 817.00 EUR
Specifications
| Antigen | Vitronectin |
|---|---|
| Clone | A18 |
| Concentration | 1 mg/mL |
| Dilution | ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500 |
| Applications | ELISA, Immunohistochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18158721
|
Novus Biologicals
NBP2-23571 |
250 μg |
817.00 EUR
250µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18126092
|
Novus Biologicals
NBP2-23571-0.05MG |
0.05 mg |
199.00 EUR
0.05mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Vitronectin Monoclonal specifically detects Vitronectin in Bovine, Equine, Rabbit samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Western Blot (Negative).Specifications
| Vitronectin | |
| 1 mg/mL | |
| ELISA, Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Immunology, Innate Immunity | |
| complement S-protein, epibolin, serum spreading factor, Serum-spreading factor, somatomedin B, S-protein, V75, vitronectin, vitronectin (serum spreading factor, somatomedin B, complement S-protein), VN, VNT | |
| VTN | |
| IgG1 κ | |
| Protein A or G purified | |
| Clone A18 is highly specific for vitronectin. There is no evidence for cross-reactivity with other connective tissue proteins (fibronectin, elastin, collagen, laminin). Clone A18 cross-reacts with rabbit and horse vitronectin, no reactivity with human or sheep. |
| A18 | |
| ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Bovine, Equine, Rabbit | |
| 7448 | |
| Lysed bovine corneal endothelial cells and extracellular matrix | |
| Primary | |
| Store at 4C. Do not freeze. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title