missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vitronectin Antibody (A18) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-23571
This item is not returnable.
View return policy
Description
Vitronectin Monoclonal specifically detects Vitronectin in Bovine, Equine, Rabbit samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Western Blot (Negative).
Specifications
| Vitronectin | |
| Monoclonal | |
| 1 mg/mL | |
| ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500 | |
| VTN | |
| Lysed bovine corneal endothelial cells and extracellular matrix | |
| 250 μg | |
| Immunology, Innate Immunity | |
| 7448 | |
| Bovine, Equine, Rabbit | |
| Purified |
| ELISA, Immunohistochemistry | |
| A18 | |
| Unconjugated | |
| complement S-protein, epibolin, serum spreading factor, Serum-spreading factor, somatomedin B, S-protein, V75, vitronectin, vitronectin (serum spreading factor, somatomedin B, complement S-protein), VN, VNT | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Clone A18 is highly specific for vitronectin. There is no evidence for cross-reactivity with other connective tissue proteins (fibronectin, elastin, collagen, laminin). Clone A18 cross-reacts with rabbit and horse vitronectin, no reactivity with human or sheep. | |
| Store at 4C. Do not freeze. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction