Learn More
ROBU™ by STEREON VitraPOR™ COD Reaction Vessel, DIN 38409
Product Code. 10594741
Change view
Click to view available options
Joint Type:
NS 29/32
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Joint Type Unit Size
10594741 NS 29/32 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
When working with glassware always wear protective glasses and protective gloves to avoid injuries. Use a safety screen, -hood or similar protection when working with glassware subject to pressure or vacuum. Be prepared that reagents may leak from a broken vessel. Please heed these guidelines in combination with the respective state-specific regulations for the use of glasware in the laboratory.
Product Code. 10594741 Supplier ROBU™ by STEREON Supplier No. 18910
  • ONLINE EXCLUSIVE PRICE
    40.75 EUR
    37.06 EUR / Pack of 1
    Save 3.69 EUR
    9% Off
 Restricted Item
Estimated Shipment: 23-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
When working with glassware always wear protective glasses and protective gloves to avoid injuries. Use a safety screen, -hood or similar protection when working with glassware subject to pressure or vacuum. Be prepared that reagents may leak from a broken vessel. Please heed these guidelines in combination with the respective state-specific regulations for the use of glasware in the laboratory.
Precisely formed borosilicate 3.3 glass for chemical oxygen demand determination in water-analysis laboratories.
  • Precisely moulded diameter supports compatibility with standardised laboratory setups
  • Standard-joint connection for use with the corresponding HCl absorber
  • Transparent glass allows direct observation of the reaction
  • Designed for environmental, wastewater and quality-control laboratories
  • Reusable glass construction for validated analytical workflows
Specifications
Product Type Reaction Vessel
Joint Type NS 29/32
Material Borosilicate Glass (3.3)
Use only within the validated temperature, reagent and analytical-method limits of the laboratory procedure.