missing translation for 'onlineSavingsMsg'
Learn More

Vitamin D3, 99.85%, MP Biomedicals™

Product Code. 11473180 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11473180 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11473180 Supplier MP Biomedicals Supplier No. 0210122401
  • ONLINE EXCLUSIVE PRICE
    68.35 EUR
    65.10 EUR / 1g
    Save 3.25 EUR
    4% Off
 Restricted Item
Estimated Shipment: 23-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 67-97-0
Assay Percent Range 0.9985
Molecular Formula C27H44O
Quantity 1 g
Synonym vitamin d3, cholecalciferol, calciol, colecalciferol, oleovitamin d3, arachitol, activated 7-dehydrocholesterol, vitamin d, colecalcipherol, colecalciferolum
InChI Key QYSXJUFSXHHAJI-YRZJJWOYSA-N
SMILES CC(C)CCCC(C)C1CCC2C1(CCCC2=CC=C3CC(CCC3=C)O)C
IUPAC Name (1S,3Z)-3-[(2E)-2-[(1R,3aS,7aR)-7a-methyl-1-[(2R)-6-methylheptan-2-yl]-2,3,3a,5,6,7-hexahydro-1H-inden-4-ylidene]ethylidene]-4-methylidenecyclohexan-1-ol
Molecular Weight (g/mol) 384.648
PubChem CID 5280795
ChEBI CHEBI:28940
Formula Weight 384.65
Physical Form Crystalline
Chemical Name or Material Vitamin D3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.