missing translation for 'onlineSavingsMsg'
Learn More

Vitamin D3, MP Biomedicals™

Product Code. 11463180 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11463180 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11463180 Supplier MP Biomedicals Supplier No. 02101224.2
  • ONLINE EXCLUSIVE PRICE
    48.50 EUR
    46.20 EUR / 250mg
    Save 2.30 EUR
    4% Off
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Is the naturally occurring form of vitamin D3. Vitamin D3, MP Biomedicals mediates intestinal calcium absorption, bone metabolism, and probably muscle activity. It functions as a hormone precursor and acts through a receptor that is a member of the ligand

It acts as a hormone precursor since it requires two stages of metabolism: first to 25-hydroxycholecalciferol; then to 1,25-dihydroxycholecalciferol.

  • Modulates the proliferation and differentiation of both normal and cancer cells
  • Has antiproliferative and antimetastatic effects on breast, colon, and prostate cancer cells
  • Activated vitamin D receptors in intestine and bone maintain calcium absorbance and homeostasis

Specifications

CAS 67-97-0
Melting Point 82°C to 92°C
Molecular Formula C27H44O
Quantity 250 mg
Synonym vitamin d3, cholecalciferol, calciol, colecalciferol, oleovitamin d3, arachitol, activated 7-dehydrocholesterol, vitamin d, colecalcipherol, colecalciferolum
Solubility Information Soluble in ethanol, acetone, ether, chloroform; insoluble in water. Practically insoluble in water: soluble in the usual organic solvents; slightly soluble in vegetable oils.
InChI Key QYSXJUFSXHHAJI-YRZJJWOYSA-N
SMILES CC(C)CCCC(C)C1CCC2C1(CCCC2=CC=C3CC(CCC3=C)O)C
IUPAC Name (1S,3Z)-3-[(2E)-2-[(1R,3aS,7aR)-7a-methyl-1-[(2R)-6-methylheptan-2-yl]-2,3,3a,5,6,7-hexahydro-1H-inden-4-ylidene]ethylidene]-4-methylidenecyclohexan-1-ol
Molecular Weight (g/mol) 384.648
PubChem CID 5280795
ChEBI CHEBI:28940
Formula Weight 384.65
Physical Form Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.