Learn More
Vitamin B12 c-lactone, TRC
Product Code. 30455263
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30455263 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30455263 Supplier Toronto Research Chemicals Supplier No. TRCV67565010MG
 Restricted Item
Estimated Shipment: 11-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 7beta,8beta-Lactocyanocobalamin
CAS 23208-66-4
Purity Grade Notes HPLC
Molecular Formula C63 H85 Co N13 O15 P
Quantity 10 mg
Synonym Cyanocobalamin Impurity F, Vitamin B12 c-lactone, 7β,8β-Lactocyanocobalamin, 7b,8b-Lactocyanocobalamin, Coα-[α-(5,6-dimethylbenzimidazolyl)]-Coβ-cyano-8β-hydroxycobamic a,b,d,e,g-pentaamide c,8-γ-lactone, Cyanocobalamin 7β,8β-lactone
InChI Formula InChI=1S/C62H87N12O15P.CN.Co/c1-29-20-37-38(21-30(29)2)74(28-69-37)56-51(83)52(39(27-75)86-56)89-90(84,85)88-31(3)26-68-47(81)17-18-58(8)36(22-45(66)79)55-61(11)59(9,24-46(67)80)35(13-15-43(64)77)50(73-61)33(5)54-60(10)25-48(82)87-62(60,19-16-44(65)78)41(71-54)23-40-57(6,7)34(12-14-42(63)76)49(70-40)32(4)53(58)72-55;1-2;/h20-21,23,28,31,34-36,39,51-52,55-56,75,83H,12-19,22,24-27H2,1-11H3,(H13,63,64,65,66,67,68,70,71,72,73,76,77,78,79,80,81,84,85);;/q;;+2/p-2/t31-,34-,35-,36+,39-,51-,52-,55-,56+,58-,59+,60-,61+,62+;;/m1../s1
SMILES C[C@@H]1CNC(=O)CC[C@]2(C)[C@@H](CC(=O)N)[C@H]3N4C2=C(C)C5=N6C(=CC7=N8C(=C(C)C9=N([C@]3(C)[C@@](C)(CC(=O)N)[C@@H]9CCC(=O)N)[Co+]468(C#N)N%10=CN([C@H]%11O[C@H](CO)[C@@H](OP(=O)([O-])O1)[C@H]%11O)c%12cc(C)c(C)cc%10%12)[C@@]%13(C)CC(=O)O[C@@]7%13CCC(=O)N)C(C)(C)[C@@H]5CCC(=O)N
Molecular Weight (g/mol) 1354.3341
Formula Weight 1353.5358
Percent Purity >95