Learn More
R&D Systems™ VisUBlock Mouse on Mouse Blocking Reagent
Product Code. 30002078
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30002078 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30002078 Supplier R&D Systems™ Supplier No. VB00101ML
  • ONLINE EXCLUSIVE PRICE
    347.00 EUR
    327.60 EUR / 1mL
    Save 19.40 EUR
    5% Off
 Restricted Item
Estimated Shipment: 04-11-2026
Blocking reagent for use with primary antibodies from mouse host and mouse tissue.
  • Reduces background staining
  • Enables staining of mouse tissue with mouse IgG primary antibodies
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store the unopened product at 2-8°C. Do not use past expiration date.
Product Line R&D Systems
Quantity 1 mL
Species Mouse