Learn More
Cole Parmer Vistek™ Vibration Isolation Platforms
Product Code. 11858651 Shop All Cole Parmer Products
Change view
Click to view available options
Load Bearing Capacity (Metric):
0 to 20 kg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Load Bearing Capacity (Metric) Unit Size
11858651 0 to 20 kg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11858651 Supplier Cole Parmer Supplier No. 1150003
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Significantly improve the performance of microscopes, lab balances, and other sensitive instruments
Cut off the transmission of vibrations from the tabletop or countertop. Effectively isolate horizontal and vertical vibration components in all six degrees of freedom.
  • Passive, mechanical and maintenance-free No air hoses or supporting hardware required
Specifications
Load Bearing Capacity (Metric) 0 to 20 kg