Learn More
SCHOTT AG Lighting and Imaging™ VisiLED Polarizer Set Only for Ring Light S40-75
Shop All Schott Products
Click to view available options
For Use With (Equipment):
VisiLED Ringlight S40-75
Unit Size:
Each
Description
- Polarization system for VisiLED Brightfield Ring Light S40-75
- Turnable polarizer with adapting thread and sliding filter as analyzer
- Polarized light by an polarizing and analyzing filter within the beam path can be used very efficiently used in Stereo Microscopy to contrast inner structure of material or to identify metal particles within others
- Not for use with any other ringllight
- To use only with VisiLED Ring Light S40-75
Specifications
Specifications
| Type | Polarization Filter |
| Description | Polarizer M90 x 1 Analyzer for 66 mm lens |
| For Use With (Application) | A Wide Variety of Tasks From Life Science to Industrial Applications |
| For Use With (Equipment) | VisiLED Ringlight S40-75 |
| Includes | Polarizer Filter and Analyzer |
| Size | M90 x 1 |