Learn More
SCHOTT AG Lighting and Imaging™ VisiLED MC-D 1100
Shop All Schott Products
Click to view available options
Voltage:
100/240 V
Unit Size:
Each
Description
- With the VisiLED Controller MC-D 1100 you can seamlessly integrate our complete portfolio of VisiLED microscopy illumination into your own environments.
- Designed for professional integration, the controller offers a robust solution to control our VisiLED light heads for stereo microscopy applications purely digital. T
- he controller comes with a comprehensive Software Development Kit (SDK), featuring an extensive library of control commands.
- Easy Integration into imaging and postprocessing software
- Create tailor-made solutions according to unique customer needs
- Complex lighting scenarios possible
- Real-time control
- Optimal contrasting thanks to broad VisiLED portfolio
- Extensive control options
Specifications
Specifications
| Type | LED Light Cube |
| Product Line | VisiLED |
| Autoclavable | Not Autoclavable |
| Dimensions (L x W x H) | 160.7 x 110.7 x 32 mm |
| For Use With (Equipment) | VisiLED Lightheads, Power supply unit and Power cord to be ordered separately |
| Voltage | 100/240 V |