Learn More
SI Analytics™ Basic Unit for Optical Detection
Product Code. 10551013 Shop All SI Analytics Products
Change view
Click to view available options
Voltage:
230 V
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Voltage Unit Size
10551013 230 V Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10551013 Supplier SI Analytics™ Supplier No. 285415709
Estimated Shipment: 09-12-2026
SI Analytics™ Basic Unit for Optical Detection includes ViscoPump II and a mini keyboard.
  • Compatable with voltages of 95V to 230V
  • Includes 1pc. ViscoPump II and mini keyboard
TRUSTED_SUSTAINABILITY
Specifications
Product Type ViscoPump II And Mini Keyboard
Voltage 230 V