Learn More
Scharlab Violet Red Bile Dextrose Agar (VRBD A.)
Product Code. 11411907
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11411907 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11411907 Supplier Scharlab Supplier No. 01295500
Scharlab Violet Red Bile Dextrose Agar is used for the enumeration of enterobacteria.
  • EP/HP/ISO/USP
  • Selective solid medium for the enumeration of enterobacteria.
  • Acccording to ISO 21528 standard and Pharmacopeial Harmonised Methods.
Specifications
Certifications/Compliance ISO 21528
Color Red
Form Solid
Media Type Violet Red Bile Dextrose Agar
Product Type Microbiology Media
Target Organism Class Gram-negative Bacteria
Formulation Yeast Extract 3.000 g/L, Pancreatic Digest of Gelatin 7.000 g/L, Bile Salts 1.500 g/L, D(+)-Glucose Monohydrate 10.000 g/L, Sodium Chloride 5.000 g/L, Neutral Red 0.030 g/L, Crystal Violet 0.002 g/L, Agar 13.000 g/L
For Use With (Application) Enumeration of Enterobacteria
Incubation Conditions 30°C to 35°C
Incubation Time 24 ±2 hr.
pH 7.4 ±0.2
Quantity 500 g
Solubility Insoluble