Learn More
Violanthrone 79, TRC
Product Code. 30456297
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30456297 1 g -
30451656 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30456297 Supplier Toronto Research Chemicals Supplier No. TRCV6345001G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 85652-50-2
Molecular Formula C50H48O4
Molecular Weight (g/mol) 712.91
Synonym 16,17-Bis(octyloxy)anthra[9,1,2-cde]benzo[rst]pentaphene-5,10-dione
SMILES CCCCCCCCOc1cc2c3ccccc3C(=O)c4ccc5c6ccc7C(=O)c8ccccc8c9cc(OCCCCCCCC)c(c1c5c24)c6c79
Specifications
Quantity 1 g
InChI Formula InChI=1S/C50H48O4/c1-3-5-7-9-11-17-27-53-41-29-39-31-19-13-15-21-35(31)49(51)37-25-23-33-34-24-26-38-44-40(32-20-14-16-22-36(32)50(38)52)30-42(54-28-18-12-10-8-6-4-2)48(46(34)44)47(41)45(33)43(37)39/h13-16,19-26,29-30H,3-12,17-18,27-28H2,1-2H3
Formula Weight 712.3553
Chemical Name or Material Violanthrone 79