Learn More
3M™ Vinyl Tape 471F
Product Code. 17207780
Change view
Click to view available options
Color:
Black
Blue
Green
Red
White
Yellow
Unit Size:
24 rolls
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
17207780 Black 24 rolls
17257770 Red 24 rolls
17267770 Blue 24 rolls
17277770 Yellow 24 rolls
17287770 White 24 rolls
17297770 Green 24 rolls
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 17207780 Supplier 3M™ Supplier No. 7000048401
  • ONLINE EXCLUSIVE PRICE
    2310.00 EUR
    1297.31 EUR / 24 rolls
    Save 1012.69 EUR
    43% Off
Estimated Shipment: 01-10-2026
Add to basket
Add to basket
Commonly used for marking hazards and aisles, color-coding, paint masking, decorative trim, splicing, and more. Made from vinyl backing with rubber adhesive, our versatile tape resists scrapes, wear, and weathering for long lasting use.
  • Maintains its color when exposed to abrasion, wear and many solvents
  • Unique stretch properties enable tape to remain stretched to conform to uneven surfaces without lifting and retracting
  • Ideal for use on curved and irregular surfaces
  • Easy to apply, excellent holding strength on many surfaces
  • Available in many distinctive colours (yellow, white, red, black, brown, green, orange, purple, blue and transparent)
  • Available in a wide variety of vivid colours and sizes
TRUSTED_SUSTAINABILITY
Specifications
Dimensions (L x W) 50 mm x 33 m
Height (Metric) 0.1 mm
Length (Metric) 32.9 m
Thickness 0.14 mm
Width (Metric) 5.1 cm
Color Black