Learn More
Brady™ Vinyl Glow-in-the-dark Anti-skid Right Footprint
Product Code. 11711965 Shop All Brady Products
Change view
Click to view available options
Color:
Photoluminescent
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
11711965 Photoluminescent Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11711965 Supplier Brady™ Supplier No. 224073
  • ONLINE EXCLUSIVE PRICE
    12.90 EUR
    12.03 EUR / Pack of 1
    Save 0.87 EUR
    6% Off
Estimated Shipment: 28-10-2026
Use this Vinyl Glow-in-the-dark Anti-skid Right Footprint together with the left foot to create a visual pathway to aid a fast exit during power outages and emergency situations. Anti-slip effect.
  • Anti-slip effect.
Specifications
Alternate Part Number GLOW IN THE DARK ANTI SKID FOOT. RIGHT
Description Glow-in-the-dark anti-skid footprint - Right foot
For Use With (Application) Use Together With the Right Foot to Create a Visual Pathway
Height (English) 8.27 in.
Height (Metric) 210 mm
Packaging Quantity 1 Ea.
Temperature Range (English) -40°F to +230°F
Temperature Range (Metric) -40°C to +110°C
Thickness 0.6 mm
Width (English) 3.35 in.
Width (Metric) 85 mm
Color Photoluminescent
Material Aluminum