Learn More
Vildagliptin-d3, TRC
Specifications
Specifications
| Chemical Name or Material | Vildagliptin-d3 |
| CAS | 1217546-82-1 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C17H22D3N3O2 |
| Quantity | 0.5 mg |
| Synonym | (2S)-1-[2-[(3-Hydroxytricyclo[3.3.1.13,7]dec-1-yl)amino]acetyl]-2-(pyrrolidine-d3)carbonitrile, LAF 237-d3, NVP-LAF 237-d3, Galvus-d3 |
| InChI Formula | InChI=1S/C17H25N3O2/c18-9-14-2-1-3-20(14)15(21)10-19-16-5-12-4-13(6-16)8-17(22,7-12)11-16/h12-14,19,22H,1-8,10-11H2/t12-,13+,14-,16?,17?/m0/s1/i3D2,14D |
| SMILES | O=C(N1[C@]([2H])(C#N)CCC1([2H])[2H])CN[C@]23C[C@](C[C@H]4C3)(O)C[C@@H](C4)C2 |
| IUPAC Name | (2S)-2,5,5-trideuterio-1-[2-[[(5R,7S)-3-hydroxy-1-adamantyl]amino]acetyl]pyrrolidine-2-carbonitrile |
| Molecular Weight (g/mol) | 306.42 |
| Show More |