Learn More
Vilazodone, TRC
Specifications
Specifications
| Chemical Name or Material | Vilazodone |
| CAS | 163521-12-8 |
| Molecular Formula | C26 H27 N5 O2 |
| Quantity | 50 mg |
| Synonym | 5-[4-[4-(5-cyano-1H-indol-3-yl)butyl]-1-piperazinyl]-2-Benzofurancarboxamide, 1-[4-(5-Cyanoindol-3-yl)butyl]-4-(2-carbamoylbenzofuran-5-yl)piperazine, EMD 515259, Vilazodone |
| InChI Formula | InChI=1S/C26H27N5O2/c27-16-18-4-6-23-22(13-18)19(17-29-23)3-1-2-8-30-9-11-31(12-10-30)21-5-7-24-20(14-21)15-25(33-24)26(28)32/h4-7,13-15,17,29H,1-3,8-12H2,(H2,28,32) |
| SMILES | N#CC1=CC2=C(NC=C2CCCCN3CCN(CC3)C4=CC5=C(OC(C(N)=O)=C5)C=C4)C=C1 |
| IUPAC Name | 5-[4-[4-(5-cyano-1H-indol-3-yl)butyl]piperazin-1-yl]-1-benzofuran-2-carboxamide |
| Molecular Weight (g/mol) | 441.52 |
| Formula Weight | 441.2165 |