Learn More
Mouse BMP-4 Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems MAB50201-SP
This item is not returnable.
View return policy
Description
BMP-4 Monoclonal specifically detects BMP-4 in Mouse samples. It is validated for Neutralization.
Specifications
| BMP-4 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BMP2B, BMP-2B, BMP2B1, BMP2BMCOPS6, BMP4, BMP-4, Bone morphogenetic protein 2B, bone morphogenetic protein 4, DVR4, MCOPS6, OFC11, ZYME | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 652 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Neutralization | |
| 871317 | |
| Neutralization 0.4-2.4 ug/mL | |
| P21275 | |
| Bmp4 | |
| Chinese hamster ovary cell line CHO-derived mature mouse BMP-4 Ser293-Arg408 Accession # P21275 | |
| 25 μg | |
| Primary | |
| Detects mouse BMP-4 in direct ELISAs. | |
| Mouse | |
| Purified |
Product Content Correction
Product Title
For Research Use Only