missing translation for 'onlineSavingsMsg'
Learn More

Mouse BMP-4 Antibody, R&D Systems™
GREENER_CHOICE

Rat Monoclonal Antibody

201.00 EUR - 396.00 EUR

Specifications

Antigen BMP-4
Clone 871317
Dilution Neutralization 0.4-2.4 ug/mL
Applications Neutralization
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18792879
GREENER_CHOICE
View Documents
R&D Systems
MAB50201-SP
25 μg
201.00 EUR
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18749907
GREENER_CHOICE
View Documents
R&D Systems
MAB50201
100 μg
396.00 EUR
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

BMP-4 Monoclonal specifically detects BMP-4 in Mouse samples. It is validated for Neutralization.
Specifications

Specifications

BMP-4
Neutralization 0.4-2.4 ug/mL
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
BMP2B, BMP-2B, BMP2B1, BMP2BMCOPS6, BMP4, BMP-4, Bone morphogenetic protein 2B, bone morphogenetic protein 4, DVR4, MCOPS6, OFC11, ZYME
Bmp4
IgG1
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
871317
Neutralization
Unconjugated
Rat
Mouse
P21275
652
Chinese hamster ovary cell line CHO-derived mature mouse BMP-4 Ser293-Arg408 Accession # P21275
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects mouse BMP-4 in direct ELISAs.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.