Learn More
Tocris Bioscience™ EN 219
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
Description
EN 219 is a covalent RNF114 inhibitor (IC50 = 470 nM). In vitro EN 219 mimics the mechanism of action of Nimbolide (Cat. No. 7289) by binding to RNF114 and inhibiting ubiquitination of its endogenous substrate, the tumor suppressor p21.
Specifications
Specifications
| Chemical Name or Material | 1-[3,5-bis(4-Bromophenyl)-4,5-dihydro-1 H-pyrazol-1-yl]-2-chloroethanone |
| CAS | 380351-29-1 |
| Quantity | 25 mg |
| Target | RNF114 |
| Molecular Formula | C17H13Br2ClN2O |
| Purity | 0.98 |