Learn More
Spectrum Labs™ Spectra/Por™ 1 Membrane Disc
Product Code. 12717526
Click to view available options
Diameter (Metric):
47 mm
100 mm
Unit Size:
Pack of 50
Product Code. 12717526
Brand: Spectrum Labs™ 132474
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Regenerated cellulose membrance for use with Spectrum Equilibrium Dializer and Spectra/Por MacroDializer systems
For use with Spectrum Equilibrium Dialyzer and Spectra/Por™ MacroDialyzer systems.
Regenerated cellulose (RC) precut membrane discs
Supplied dry with a glycerol as a humectant
Specifications
Product Type Membrane Disc
MWCO 6 to 8 kDa
Membrane Regenerated Cellulose
Diameter (Metric) 100 mm
For Use With (Application) For Basic Dialysis