Learn More
Thermo Scientific™ Sterilin™ Polypropylene 30mL Universal Containers
Description
Secure excellent temperature and chemical resistance with these 95kPa compliant 30mL leak-resistant Thermo Scientific™ Sterilin™ Polypropylene 30mL Universal Containers.
NOT AVAILABLE IN AMERICA
Features:
- Aseptically manufactured, ultra clear polypropylene
- Leak-tested in accordance with BS EN 14254 Annexe D
- Suitable for centrifugation at 9,500 × g
- Multi-seal cap 'QuickStart' for improved handling and unrivaled leak-free performance
- Lot number printed on each container to aid full traceability
- Labeled for patient details and available either undosed, dosed with boric acid preservative (0.4g) or a loose spoon
Ordering Alerts:
Not available in all geographic regions
Specifications
Specifications
| Product Type | Container |
| Material | Polypropylene |
| Shape | Cylindrical |
| Sterility | Sterile |
| Capacity (Metric) | 30 mL |
| Certifications/Compliance | Leak-tested in accordance with BS EN 14254 Annexe. |
| Closure Type | Multi-Seal |
| Description | Universal Container with Spoon |
| Diameter (Metric) | 31 mm |
| Height (Metric) | 94 mm |
| Show More |