Learn More
Lonza Pyrogen-free Test Tubes
Product Code. 11600311
Change view
Click to view available options
Length (Metric):
75 mm
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize
11600311 75 mm Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11600311 Supplier Lonza Supplier No. N201
Add to basket
Add to basket
Recommended for use as reaction tubes in gel clot assays. Lonza™ Pyrogen-free Test Tubes Product are made from USP Type 1 flint borosilicate glass and contain less than 0.005 EU/ml endotoxin.
  • Made from USP Type 1 flint borosilicate glass
  • Contain less than 0.005 EU/ml endotoxin
  • Recommended for use as reaction tubes in gel clot assays.
TRUSTED_SUSTAINABILITY
Specifications
Length (Metric) 75 mm
Material Borosilicate Glass
Closure Type Screw Cap
Product Type Test Tube