Learn More
Lead(II) 2-ethylhexanoate
Product Code. 11394237
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
Product Code. 11394237
Brand: Thermo Scientific Alfa Aesar 036711.30
Request Bulk or Custom Format
CAS: 301-08-6 | C16H30O4Pb | 493.612 g/mol
Films of lead (II) 2-ethylhexanote can be cast on silicon solutions by spin coating. Lead (II) 2-ethylhexanote is a common solvent in metal-organic deposition (MOD) process. Intermetallic PtPb nanoparticles have been synthesized by the chemical reduction of dimethyl(1,5-cyclooctadiene)platinum and lead(II)2-ethylhexanoate by sodium naphthalide in THF or diglyme. The precursors used for the photochemical production of PZT or PLZT films were lead(II) 2-ethylhexanoate, lanthanum(III) 2-ethylhexanoate, zirconyl(IV) 2-ethylhexa- noate, and titanium(IV) isopropoxide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 301-08-6
Molecular Formula C16H30O4Pb
Molecular Weight (g/mol) 493.612
MDL Number MFCD00014003
InChI Key RUCPTXWJYHGABR-UHFFFAOYSA-L
Synonym lead bis 2-ethylhexanoate, lead ii 2-ethylhexanoate, hexanoic acid, 2-ethyl-, lead salt, hexanoic acid, 2-ethyl-, lead salt 1:?, lead tallate 2-ethylhexanoate, lead 2+ bis 2-ethylhexanoate, cis-9-octadecenoamide, plumbous 2-ethylhexanoate, acmc-1aj1i, 2-ethylhexanoate; lead 2+
PubChem CID 160451
IUPAC Name 2-ethylhexanoate;lead(2+)
SMILES CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Pb+2]
Specifications
Density 1.56 g/mL
Odor Odorless
Quantity 250 g
UN Number UN3082
Solubility Information Insoluble in water.
Formula Weight 493.61
Physical Form Viscous Liquid
Chemical Name or Material Lead(II) 2-ethylhexanoate
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lead(II) 2-ethylhexanoate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Lead(II) 2-ethylhexanoate
RUO – Research Use Only