Learn More
GFL Gesellschaft Fuer Labortec™ Stainless Steel Single Distillation
Product Code. 10394441
Change view
Click to view available options
Flow Rate:
2 L/hr.
4 L/hr.
8 L/hr.
Height (Metric):
410 mm
420 mm
460 mm
490 mm
540 mm
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Flow Rate Height (Metric) unitSize
10394441 2 L/hr. 540 mm Each
10181271 4 L/hr. 490 mm Each
10079580 2 L/hr. 420 mm Each
10737071 4 L/hr. 460 mm Each
10089580 2 L/hr. 410 mm Each
10140991 8 L/hr. 540 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 10394441 Supplier GFL Gesellschaft Fuer Labortec™ Supplier No. 2008
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Use these stills for a variety of distillation applications. GFL Gesellschaft fuer Labortec™ Stainless Steel Single Distillation stills produce ultra-pure, low-gas, bacteria and pyrogen free distillate with a very low conductivity.
  • Good distillate quality, conductivity approx. 2.3μs/cm at 25°C
  • Heating element made of stainless steel, material no. 1.4876
  • Thermostatic low water cut-off, to protect the heating element in case of low water
  • Energy-saving through distillation of the heated cooling water
  • Degassing of carbon dioxide through vent in the top evaporator with baffle is easily accessible by lifting the lids.
  • Storage tank for distillate accepts double the hourly capacity of the unit
  • Condenser (cooling coil) in the storage tank, easily exchangeable
  • Water supply through a built-in solenoid valve with connection for water pressure hose 1/2 inch (inner
  • Ø 12.7mm)
  • Cooling water pressure required: >3 bar to max. 7 bar
  • After switching on the main switch the solenoid valve opens the water supply and closes it once the storage tank is full, thus avoiding unnecessary waste of water
  • cooling water outlet with hose connection 3/4 inch (inner Ø 19mm)
  • Water that has not been condensed flows off through the cooling water outlet
  • Distillate withdrawal through the drain cock on the front of the unit
  • The drain cock can be opened in either continuous or touch position
  • An electronic level switch switches the still off when the storage tank is full and restarts it automatically when distillate is withdrawn
  • Electronic impurity detector switches the unit off in case of high degree impurities in the evaporator, the red pilot lamp "Clean" will glow
  • Drainage of the evaporator through drain cock on the right-hand side of the unit
  • Main switch and pilot lamps (yellow pilot lamp for operation and red pilot lamp for cleaning) are on the front of the unit
  • Double-walled housing. Housing is made of electrolytically galvanized sheet steel, electrostatically powder-coated with epoxy resin
  • Power connection through connection cable; 2 and 4 liter units have German shock-proof type (Schuko) plug
Specifications
Type Single Distillation
Frequency 60 Hz
Voltage 230/50 V
Flow Rate 2 L/hr.
Depth (Metric) 410 mm
Height (Metric) 540 mm
Width (Metric) 780 mm