missing translation for 'onlineSavingsMsg'
Learn More

VI 16832, MedChemExpress

Product Code. 16490018
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16490018 5 mg 5mg
16477486 100 mg 100mg
16461739 1 mg 1mg
16423567 50 mg 50mg
16407548 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16490018 Supplier Medchem Express Supplier No. HY186235mg
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress VI 16832 is a broad spectrum Type I kinase inhibitor which can be used as an enrichment tool for the comparative expression analysis of protein kinases in different cancer cell lines.

VI 16832 is a broad spectrum Type I kinase inhibitor which can be used as an enrichment tool for the comparative expression analysis of protein kinases in different cancer cell lines.

Specifications

Chemical Name or Material VI 16832
CAS 1430218-51-1
Color Yellow
Purity Grade Notes Research
Molecular Formula C22H25N5O2
Quantity 5 mg
Solubility Information DMSO : 33.33 mg/mL (85.14 mM; Need ultrasonic)
SMILES O=C1C=CC2=CN=C(NC3=CC=C(OCCN)C=C3)N=C2N1C4C(C5)CCC5C4
Molecular Weight (g/mol) 391.47
Formula Weight 391.47
Percent Purity 99.72%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.