missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ VH 101, phenol

Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Description
VH 101, phenol is a functionalized von-Hippel-Lindau protein ligand (VHL) for PROTAC® research and development; incorporates an E3 ligase ligand with terminal hydroxyl ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D.
Please contact us for SD files of our available Degrader Building Blocks.
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifications
Specifications
| Chemical Name or Material | (2 S,4 R)-1-(-2-(1-Fluorocyclopropane-1-carboxamido)-3,3-dimethylbutanoyl)-4-hydroxy-N-(2-hydroxy-4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide |
| CAS | 2306193-99-5 |
| Quantity | 50 mg |
| Target | Degrader Building Blocks |
| Molecular Formula | C26H33FN4O5S |
| Formula Weight | 6414 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction