Learn More
Tocris Bioscience™ VH 032 amide-alkylC5-acid
Description
VH 032 amide-alkylC5-acid is a functionalized von-Hippel-Lindau protein ligand (VHL) for PROTAC™ research and development; incorporates an E3 ligase ligand plus alkylC5 linker with terminal carboxylic acid ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D. Please contact us for SD files of our available Degrader Building Blocks.PROTAC™ is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifications
Specifications
| Chemical Name or Material | 7-(((S)-1-((2S,4R)-4-Hydroxy-2-((4-(4-methylthiazol-5-yl)benzyl)carbamoyl)pyrrolidin-1-yl)-3,3-dimethyl-1-oxobutan-2-yl)amino)-7-oxoheptanoic acid |
| CAS | 2162120-87-6 |
| Quantity | 25 mg |
| Target | Degrader Building Blocks |
| Molecular Formula | C29H40N4O6S |
| Purity | 95% |