Learn More
Ansell™ VersaTouch™ 23-201 Series Blue PVC Gloves
Product Code. 15361005 Shop All Ansell Products
Change view
Click to view available options
Size:
7
8
9
10
Unit Size:
6 pairs
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15361005 10 6 pairs
15341005 8 6 pairs
15331005 7 6 pairs
15351005 9 6 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15361005 Supplier Ansell™ Supplier No. 23201100
  • ONLINE EXCLUSIVE PRICE
    189.60 EUR
    123.84 EUR / 6 pairs
    Save 65.76 EUR
    34% Off
Estimated Shipment: 07-01-2027
Comfort long sleeve PVC glove with good dexterity and pliability even at low temperature.
  • Rough sandblast finish provide good wet-grip
  • Soft PVC coating on glove maintains dexterity and pliabiity at low temperatures
  • Dipped rough finish makes handling wet and slippery materials easier and safer
  • Heat sealed sleeve offers liquid resistance and dust protection only
Applications:
  • Chemical handling
  • Chemical refining
  • Cleaning
  • Construction work
  • Dairy processing
  • Fish processing
  • Fruit and vegetable processing
  • General purpose
  • Janitorial use
  • Light assembly tasks
  • Maintenance
  • Refining oil and petrol
TRUSTED_SUSTAINABILITY
Specifications
Back Material PVC
Certifications/Compliance Standards CATEGORY III, CE Mark, EN 374, EN 388 : 4121
Cuff Material PVC
Applications Industrial
Coating Material PVC
Color Blue
Latex Free Yes
Material PVC
Product Type Chemical-resistant Gloves
Size 10
Thickness (Metric) 4 mm
Glove Cuff Style Heat-sealed Sleeve
Lining Cotton
Quantity 72 Pairs
Resistant To Chemical and liquid
Series 23-201