Learn More
3M™ Versaflo™ Premium Replacement Hood S-657 with Inner Shroud
Product Code. 30690001 Shop All 3M Products
Change view
Click to view available options
Product Type:
Hood Suspension
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
30690001 Hood Suspension Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30690001 Supplier 3M™ Supplier No. 7100350374
  • ONLINE EXCLUSIVE PRICE
    695.00 EUR
    632.47 EUR / Pack of 10
    Save 62.53 EUR
    8% Off
Estimated Shipment: 07-12-2026
Offers a variety of options for faceseals, collars, shrouds and suspensions, allowing you to choose the respiratory protection
  • Lightweight, versatile and easy to maintain
  • 3M™ Versaflo™ S-Series Headcovers and Hoods help protect your workers from a variety of contaminants while maintaining comfort
  • Convenience of fully assembled integrated hoods or the cost-savings of a reusable suspension
TRUSTED_SUSTAINABILITY
Specifications
Color White
Disposable Reusable
Material Plastic
Product Type Hood Suspension
Size One Size
With Exhalation Valve No