Learn More
3M™ Versaflo™ PAPR Breathing Tube Covers
Product Code. 17752908 Shop All 3M Products
Change view
Click to view available options
For Use With (Equipment):
TR-300+ PAPR
TR-600/800 PAPR
Unit Size:
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
17752908 TR-300+ PAPR Pack of 10
17742908 TR-600/800 PAPR Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17752908 Supplier 3M™ Supplier No. 7100260719
  • ONLINE EXCLUSIVE PRICE
    343.00 EUR
    244.72 EUR / Pack of 10
    Save 98.28 EUR
    28% Off
View all promotions
Estimated Shipment: 14-01-2027
PAPR disposable breathing covers are designed to help keep Versaflo™ PAPRs clean from debris.
  • Made from a liquid resistant material with zippers for convenient donning and doffing
  • Clear plastic window for control viewing and button activation
  • Helps keep PAPR clean from debris
  • Compatible with Versaflo™ PAPRs
  • Popular for paint spray application
  • Made with no natural rubber latex components
TRUSTED_SUSTAINABILITY
Specifications
Material Plastic
Product Type Breathing Tube Cover
For Use With (Application) Protects PAPR blower from dirt and debris
For Use With (Equipment) TR-300+ PAPR
Model TR-381
Quantity 10/Cs.