Learn More
3M™ Versaflo™ M-Series Faceshields and Helmets
Product Code. 17825666 Shop All 3M Products
Change view
Click to view available options
Includes:
Comfort Faceseal
Flame Resistant Faceseal
Flame Resistant Poly Faceseal
Flame Resistant Shroud
Highly Durable Shroud
Color:
Black
Light Gray
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Color Unit Size
17825666 Comfort Faceseal Light Gray Each
17855666 Flame Resistant Poly Faceseal Light Gray Each
17835666 Comfort Faceseal Black Each
17809271 Flame Resistant Faceseal Black Each
11783916 Highly Durable Shroud Black Each
17815666 Flame Resistant Shroud Black Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 17825666 Supplier 3M™ Supplier No. 7100258667
  • ONLINE EXCLUSIVE PRICE
    357.00 EUR
    258.88 EUR / Pack of 1
    Save 98.12 EUR
    27% Off
Estimated Shipment: 19-10-2026
3M™ Versaflo™ M-Series Faceshields and Helmets can help protect your workers from a variety of respiratory and impact hazards while maintaining comfort. They are are lightweight, compact and easy to maintain.
  • M-Series are specifically designed to work together with your 3M™ Versaflo™ Powered Air Respirator and 3M™ Supplied Air Respirator Systems.
  • Deflector allows users to direct the airflow inside the headtop to increase comfort.
  • Fully-adjustable suspension for customized fit and weight distribution.
  • Accessory options availalble for protection to help handle the environments you encounter.
  • Visor combines good peripheral and downward vision.
  • Specific headtops can be used with a range of 3M™ Peltor™ hearing protection as well as an attachable microphone for ease of communication.
TRUSTED_SUSTAINABILITY
Specifications
Color Light Gray
Design Type Comfort Faceseal
Disposable Reusable
Filter Type None
Material Polyethylene
Product Type Hood with Faceseal
Size Standard
With Exhalation Valve No
Includes Comfort Faceseal