Learn More
Unity Lab Services™ Verification Report Burette
Description
- Including: external cleaning
- Verification of volumes delivered in three volumes, five weighings
- Judgment according to criteria of NF EN ISO 8655-3
- Drafting of the verification report
- Average time: 15 days from receipt of material and order
Specifications
Specifications
| For Use With (Equipment) | Burettes |
| Product Type | Verification Report |